Läs mer
AM-0902, MedChemExpress
Produktkod. 16486897
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16486897 5 mg 5mg
16425146 100 mg -
16414188 10 mg -
16410537 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16486897 Leverantör Medchem Express Leverantörsnummer HY108329
MedChemExpress AM-0902 is a potent, selective transient receptor potential A1 (TRPA1) antagonist with IC50s of 71 and 131 nM for rTRPA1 and hTRPA1, respectively.
AM-0902 is a potent, selective transient receptor potential A1 (TRPA1) antagonist with IC50s of 71 and 131 nM for rTRPA1 and hTRPA1, respectively.
Specifikationer
Kemiskt namn eller material AM-0902
CAS 1883711-97-4
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C17H15ClN6O2
Kvantitet 5 mg
Löslighetsinformation DMSO : 150 mg/mL (404.54 mM; Need ultrasonic and warming)
LEDER O=C1N(CC2=NC(CCC3=CC=C(Cl)C=C3)=NO2)C=NC4=C1N(C)C=N4
Molekylvikt (g/mol) 370.79
Formel vikt 370.79
Procent renhet 99.67%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation