Läs mer
AM 0902, Tocris Bioscience™
Potent and selective TRPA1 antagonist
Brand: Tocris Bioscience 5914/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
AM 0902 is a potent and selective TRPA1 antagonist (IC50 = 0.02 μM for human TRPA1). Exhibits no siginificant activity against human TRPV1 or TRPV4, and rat TRPV1, TRPV3 or TRPM8. Exhibits efficacy in a rat pain model. Orally bioavailable.Specifikationer
| 1-[[3-[2-(4-Chlorophenyl)ethyl]-1,2,4-oxadiazol-5-yl]methyl]-1,7-dihydro-7-methyl-6H-purin-6-one | |
| 1883711-97-4 | |
| >98% | |
| 10 mg |