missing translation for 'onlineSavingsMsg'
Läs mer

AM-2394, MedChemExpress

Produktkod. 16479506
Encompass_Preferred
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16479506 50 mg
16435597 5 mg
16434266 100 mg
16412858 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16479506 Leverantör Medchem Express Leverantörsnummer HY100221

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

MedChemExpress AM-2394 is a structurally distinct glucokinase activator (GKA). AM-2394 activates glucokinase (GK) with an EC50 of 60 nM.

AM-2394 is a structurally distinct glucokinase activator (GKA). AM-2394 activates glucokinase (GK) with an EC50 of 60 nM.

Specifikationer

Kemiskt namn eller material AM-2394
CAS 1442684-77-6
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C22H25N5O4
Kvantitet 50 mg
Löslighetsinformation DMSO : ≥ 30 mg/mL (70.84 mM)
LEDER O=C(NC)NC1=CC(OC2=CC(C)=CN=C2)=C(C3=CC=C(OCC(C)(O)C)N=C3)C=N1
Molekylvikt (g/mol) 423.47
Formel vikt 423.47
Procent renhet 99.48%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Metabolism-sugar/lipid metabolism
Visa mer Visa mindre

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.