Läs mer
Tocris Bioscience™ AM 6545
High affinity and selective CB1 antagonist
Brand: Tocris Bioscience™ 5443/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
AM 6545 is a high affinity and selective CB1 antagonist (Ki = 3.3 nM). Exhibits >100-fold selectivity for CB1 over CB2 receptors. Improves glucose homeostasis, reverses hepatic steatosis and reduces body weight in diet-induced obese mice. Orally available and non-brain penetrant.Specifikationer
| AM 6545 | |
| 5-[4-(4-Cyano-1-butyn-1-yl)phenyl]-1-(2,4-dichlorophenyl)-N-(1,1-dioxido-4-thiomorpholinyl)-4-methyl-1H-pyrazole-3-carboxamide | |
| Soluble to 20mM in DMSO | |
| Bioactive Small Molecule |
| 1245626-05-4 | |
| 10 mg | |
| Solid |
Produkttitel
Research Use Only. Not For Human or Veterinary Use