Läs mer
AM095, MedChemExpress
Produktkod. 16496968
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Förpackningsstorlek:
100mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16496968 10 mg -
16463027 50 mg -
16457046 100 mg 100mg
16449517 5 mg -
16431216 200 mg -
16401806 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Produktkod. 16496968 Leverantör Medchem Express Leverantörsnummer HY16039
MedChemExpress AM095 is a selective LPA1 receptor antagonist. The IC50 for AM095 antagonism of LPA-induced calcium flux of human or mouse LPA1-transfected CHO cells is 0.025 and 0.023 μM, respectively.
AM095 is a selective LPA1 receptor antagonist. The IC50 for AM095 antagonism of LPA-induced calcium flux of human or mouse LPA1-transfected CHO cells is 0.025 and 0.023 μM, respectively.
Specifikationer
Kemiskt namn eller material AM095
Fysisk form Solid
CAS 1345614-59-6
Färg Earth Yellow
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C27H23N2NaO5
För användning med (applikation) COVID-19-immunoregulation
Löslighetsinformation DMSO : 83.33 mg/mL (174.16 mM; Need ultrasonic)
LEDER O=C(O[Na])CC1=CC=C(C2=CC=C(C3=C(NC(O[C@@H](C4=CC=CC=C4)C)=O)C(C)=NO3)C=C2)C=C1
Molekylvikt (g/mol) 478.47
Formel vikt 478.47
Procent renhet 98.0%