Läs mer
Tocris Bioscience™ α-Amanitin
Beskrivning
Inhibitor of RNA polymerase II. Inhibits transcription in eukaryotic cells. Binds and blocks the largest subunits of RNA polymerase II, preventing new ribonucleotides from incorporating into the nascent RNA chain. Potent amatoxin.
Specifikationer
Specifikationer
| CAS | 23109-05-9 |
| Kvantitet | 1 mg |
| Produkttyp | α-Amanitin |
| Renhet | >95% |
| Molekylvikt (g/mol) | 918.977 |