Läs mer
Ambenonium Dichloride, TRC
Produktkod. 18083310
Change view
Kvantitet:
250 mg
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
18083310 2.5 g 2.5g
18091110 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18083310 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA5758752.5G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 115-79-7
Molekylformel C28 H42 Cl2 N4 O2 . 2 Cl
Synonym Benzenemethanaminium, N,N'-[(1,2-dioxo-1,2-ethanediyl)bis(imino-2,1-ethanediyl)]bis[2-chloro-N,N-diethyl-, chloride (1:2)∣Ammonium, [oxalylbis(iminoethylene)]bis[(o-chlorobenzyl)diethyl-, dichloride (8CI)∣Benzenemethanaminium, N,N'-[(1,2-dioxo-1,2-ethanediyl)bis(imino-2,1-ethanediyl)]bis[2-chloro-N,N-diethyl-, dichloride (9CI)∣[Oxalylbis(iminoethylene)]bis[(o-chlorobenzyl)diethylammonium chloride] (6CI,7CI)∣Ambenonium chloride∣Ambenonium dichloride∣Ambestigmin chloride∣Misuran∣Mysuran∣Mysuran chloride∣Mytelase∣Mytelase chloride∣N,N'-Bis-(2-diethylaminoethyl)oxamide bis-2-chlorobenzylchloride∣N,N'-Bis-2-[(2-chlorobenzyl)diethylammonium chloride]-ethyloxamide∣Oxamizil∣Oxazil∣Oxazyl∣Win 8077
Specifikationer
Kemiskt namn eller material Ambenonium Chloride
Kvantitet 2.5 g
Frakt skick Room Temperature
Formel vikt 606.2062
Molekylvikt (g/mol) 608.47