missing translation for 'onlineSavingsMsg'
Learn More

Amberlite 200, MP Biomedicals™

Catalog Number p-4774254
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
12626-25-4
MFCD00145784
This item is not returnable. View return policy
12626-25-4
MFCD00145784

Strongly acidic macroreticular-type cation exchange resin. Sodium form sulfonic acid functionality

Physical Form Powder
Color Yellow
Quantity 100 g
Chemical Name or Material Amberlite
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'