missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNAqueous™-Midi Total RNA Isolation Kit

RNA isolation for midrange amounts of starting material

Brand:  Invitrogen™ AM1911

Additional Details : Weight : 1.08000kg

Product Code. 10015954

  • 3350.00 SEK / Each


Estimated Shipment date
from Supplier 12-06-2020
Add to basket

For Research Use Only. All usage must comply with product instructions.


Description

Description

The Ambion RNAqueous-Midi Kit is for phenol-free total RNA isolation from 0.1g to 0.5g of tissue or 107 to 108 cells using a guanidinium-based lysis/;denaturant and glass fiber filter separation technology. The RNAqueous-Midi Kit provides enough reagents for 15 RNA purifications. The Ambion RNAqueous-Midi Kit uses a powerful chaotropic solution for cell lysis, followed by RNA binding to glass fiber housed in a luer-lock syringe filter. Washing and elution solutions are simply pushed through the filter using a standard syringe (not included). Vacuum filtration is not required; however, the filter units are compatible with standard vacuum manifolds for processing several samples at once.

  • Convenient syringe-based filtration
  • Compatible with vacuum manifolds when processing multiple samples

Not high-throughput compatible (Manual)

Applications:

DNA and RNA Purification and Analysis, RNA Extraction, Total RNA Isolation, Total RNA from Animal Cells and Tissues

Specifications

Specifications

Total RNA
Total RNA
Formaldehyde Load Dye, Lysis/Binding Solution, Water for 64% Ethanol, Lithium, Chloride Solution, Wash Solution No. 1 and Wash Solution 2/3 Concentrate, Syringe Filter Units, Collection Tubes and Elution Solution
Kit
Not High-throughput Compatible (Manual)
Ambion™, RNAqueous®
Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Nuclease Protection Assays, Northern Blotting, cDNA Library Construction
Tissue, Cells
Syringe Filter (Glass Fiber)

Formaldehyde Load Dye should bestored at -20°C; Lysis/Binding Solution, Water for 64% Ethanol, LithiumChloride Solution, Wash Solution #1 and Wash Solution 2/3 Concentrate should bestored at 4°C; Syringe Filter Units, Collection Tubes and Elution Solution canbe stored at room temperature. All reagents are guaranteed stable for 6 months whenproperly stored.
Room Temperature
Videos
Documents