Läs mer
Ambrisentan Acyl Beta-D-Glucuronide, TRC
Produktkod. 30316398
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30316398 1 mg
1 options
Produktkod. 30316398 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA5758701MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ambrisentan Acyl beta-D-Glucuronide
CAS 1106685-58-8
Molekylformel C28H30N2O10
Kvantitet 1 mg
Synonym 1-[(αS)-α-[(4,6-Dimethyl-2-pyrimidinyl)oxy]-β-methoxy-β-phenylbenzenepropanoate] β-D-Glucopyranuronic Acid
InChI formel InChI=1S/C28H30N2O10/c1-15-14-16(2)30-27(29-15)39-23(25(36)40-26-21(33)19(31)20(32)22(38-26)24(34)35)28(37-3,17-10-6-4-7-11-17)18-12-8-5-9-13-18/h4-14,19-23,26,31-33H,1-3H3,(H,34,35)/t19-,20-,21+,22-,23+,26-/m0/s1
LEDER CC1=CC(=NC(=N1)O[C@H](C(=O)O[C@H]2[C@@H]([C@H]([C@@H]([C@H](O2)C(=O)O)O)O)O)C(C3=CC=CC=C3)(C4=CC=CC=C4)OC)C
IUPAC-namn (2S,3S,4S,5R,6S)-6-[(2S)-2-(4,6-dimethylpyrimidin-2-yl)oxy-3-methoxy-3,3-diphenylpropanoyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
Molekylvikt (g/mol) 554.55