Läs mer
Ambrisentan Methyl Ester, TRC
Produktkod. 30135026
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135026 1 g
30136903 500 mg
30040552 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135026 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA5758711G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1106685-61-3
Molekylformel C23H24N2O4
Molekylvikt (g/mol) 392.45
Synonym (+)-(2S)-2-[(4,6-Dimethylpyrimidin-2-yl)oxy]-3-methoxy-3,3-diphenylpropanoic Acid Methyl Ester, (S)-Methyl 2-((4,6-Dimethylpyrimidin-2-yl)-oxy)-3-methoxy-3,3-diphenylpropanoate;
LEDER COC(=O)[C@@H](Oc1nc(C)cc(C)n1)C(OC)(c2ccccc2)c3ccccc3
Specifikationer
Kemiskt namn eller material Ambrisentan Methyl Ester
Kvantitet 1 g
Formel vikt 392.1736