Läs mer
Ambroxol Cycloimine Impurity, TRC
Produktkod. 30137566
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137566 25 mg
30010110 10 mg
30006702 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137566 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD42597025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1797894-71-3
Molekylformel C14 H16 Br2 N2 O
Molekylvikt (g/mol) 388.1
Synonym 4-(6,8-Dibromo-3(4H)-quinazolinyl)-cyclohexanol, 4-(6,8-dibromo-3(4H)-quinazolinyl)cyclohexanol
IUPAC-namn 4-(6,8-dibromo-4H-quinazolin-3-yl)cyclohexan-1-ol
LEDER OC1CCC(CC1)N2Cc3cc(Br)cc(Br)c3N=C2
Specifikationer
Kemiskt namn eller material 4-(6,8-Dibromo-3(4H)-quinazolinyl)-cyclohexanol
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C14H16Br2N2O/c15-10-5-9-7-18(8-17-14(9)13(16)6-10)11-1-3-12(19)4-2-11/h5-6,8,11-12,19H,1-4,7H2
Formel vikt 385.9629
Procent renhet >95%