Learn More
Ambroxol Hydrochloride, TRC
Product Code. 30134007
Change view
Click to view available options
Quantity:
100 mg
1 g
25 g
Unit Size:
100mg
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134007 25 g 25g
17994085 1 g 1g
17930755 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134007 Supplier Toronto Research Chemicals Supplier No. TRCA57590025G
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 23828-92-4
Molecular Formula C13 H18 Br2 N2 O . Cl H
Molecular Weight (g/mol) 414.56
Synonym Cyclohexanol, 4-[(2-amino-3,5-dibromobenzyl)amino]-, hydrochloride, trans- (8CI), Cyclohexanol, 4-[[(2-amino-3,5-dibromophenyl)methyl]amino]-, monohydrochloride, trans- (9CI), Abramen, Ambril, Ambrodil, Ambron, Ambroxol hydrochloride, Bronchopront, Coughnol, Duramucal, Fluibron, Fluixol, Frenopect, Lindoxyl, Motosol, Mucasan, Muco-Burg, Mucoclear, Mucofar, Mucolear, Mucolin, Mucolite, Mucosal, Mucosolvan, Mucovent, N-(trans-4-Hydroxycyclohexyl)-2-amino-3,5-dibromobenzylamine hydrochloride, Pect, Solvolan, Stas-Hustenloser, Surbronc, Surfactal, trans-4-[(2-Amino-3,5-dibromobenzyl)amino]cyclohexanol hydrochloride
IUPAC Name 4-[(2-amino-3,5-dibromophenyl)methylamino]cyclohexan-1-ol;hydrochloride
SMILES Cl.Nc1c(Br)cc(Br)cc1CN[C@@H]2CC[C@@H](O)CC2
Specifications
Quantity 25 g
InChI Formula InChI=1S/C13H18Br2N2O.ClH/c14-9-5-8(13(16)12(15)6-9)7-17-10-1-3-11(18)4-2-10;/h5-6,10-11,17-18H,1-4,7,16H2;1H/t10-,11-;
Formula Weight 411.9553
Percent Purity >95%
Chemical Name or Material Ambroxol hydrochloride
Grade HPLC