Läs mer
Ambucetamide, TRC
Produktkod. 30326682
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30326682 50 mg
1 options
Produktkod. 30326682 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA57593550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ambucetamide
CAS 519-88-0
Molekylformel C17 H28 N2 O2
Kvantitet 50 mg
Synonym Acetamide, 2-(dibutylamino)-2-(p-methoxyphenyl)- (7CI,8CI), (+/-)-Ambucetamide, 2-(Di-n-butylamino)-2-(4-methoxyphenyl)acetamide, 2-(Di-n-butylamino)-2-(p-methoxyphenyl)acetamide, 2-(Dibutylamino)-2-(p-methoxyphenyl)acetamide, A 16, Ambucetamide, Bersen, Dibutamide, Meritin, R 5, alpha-Dibutylamino-alpha-(p-methoxyphenyl)acetamide, alpha-p-Methoxyphenyl-alpha-di-n-butylaminoacetamide, Benzeneacetamide, alpha-(dibutylamino)-4-methoxy-
InChI formel InChI=1S/C17H28N2O2/c1-4-6-12-19(13-7-5-2)16(17(18)20)14-8-10-15(21-3)11-9-14/h8-11,16H,4-7,12-13H2,1-3H3,(H2,18,20)
LEDER CCCCN(CCCC)C(C(=O)N)c1ccc(OC)cc1
IUPAC-namn 2-(dibutylamino)-2-(4-methoxyphenyl)acetamide
Molekylvikt (g/mol) 292.42
Formel vikt 292.2151