Läs mer
Amdinocillin (>85%), TRC
Produktkod. 30324076
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30324076 1 g
1 options
Produktkod. 30324076 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA5764051G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Mecillinam
CAS 32887-01-7
Molekylformel C15 H23 N3 O3 S
Kvantitet 1 g
Synonym 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[(hexahydro-1H-azepin-1-yl)methylene]amino]-3,3-dimethyl-7-oxo-, (+)- (8CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[(hexahydro-1H-azepin-1-yl)methylene]amino]-3,3-dimethyl-7-oxo-, [2S-(2α,5α,6β)]-, (2S,5R,6R)-6-[[(Hexahydro-1H-azepin-1-yl)methylene]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[(Hexahydro-1H-azepin-1-yl)methylene]amino]penicillanic acid, 6β-[(Hexahydro-1H-azepin-1-yl)methyleneamino]penicillanic acid, Amdinocillin, Coactin, FL 1060, Hexacillin, Hexapen, Mecillinam, Penicillin Hx
InChI formel InChI=1S/C15H23N3O3S/c1-15(2)11(14(20)21)18-12(19)10(13(18)22-15)16-9-17-7-5-3-4-6-8-17/h9-11,13H,3-8H2,1-2H3,(H,20,21)/b16-9+/t10-,11+,13-/m1/s1
LEDER CC1(C)S[C@@H]2[C@H](\N=C\N3CCCCCC3)C(=O)N2[C@H]1C(=O)O
IUPAC-namn (2S,5R,6R)-6-[(E)-azepan-1-ylmethylideneamino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Molekylvikt (g/mol) 325.43
Formel vikt 325.146