Läs mer
Amebucort, MedChemExpress
Produktkod. 16492518
Encompass_Preferred
Change view
Kvantitet:
1 mg
5 mg
10 mg
20 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16492518 5 mg
16480319 10 mg
16463309 1 mg
16400826 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16492518 Leverantör Medchem Express Leverantörsnummer HYU00298
MedChemExpress Amebucort is a synthetic glucocorticoid corticosteroid, may used for the research of inflammatory disorders.
Amebucort is a synthetic glucocorticoid corticosteroid, may used for the research of inflammatory disorders.
Specifikationer
Kemiskt namn eller material Amebucort
CAS 83625-35-8
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C28H40O7
Kvantitet 5 mg
Löslighetsinformation DMSO : 100 mg/mL (204.66 mM; Need ultrasonic)
LEDER C[C@@]12[C@](C(COC(C)=O)=O)(OC(CCC)=O)CC[C@@]1([H])[C@]3([H])C[C@H](C)C4=CC(CC[C@]4(C)[C@@]3([H])[C@@H](O)C2)=O
Molekylvikt (g/mol) 488.61
Formel vikt 488.61
Procent renhet 98.04%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation