Läs mer
Amentoflavone, TRC
Produktkod. 17990005
Change view
Kvantitet:
1 mg
2.5 mg
100 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
17990005 1 mg -
17947374 100 mg -
17979184 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17990005 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA5764201MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1617-53-4
Molekylformel C30 H18 O10
Molekylvikt (g/mol) 538.46
Synonym 4H-1-Benzopyran-4-one, 8-[5-(5,7-dihydroxy-4-oxo-4H-1-benzopyran-2-yl)-2-hydroxyphenyl]-5,7-dihydroxy-2-(4-hydroxyphenyl)-, 3' '',8-Biflavone, 4',4' '',5,5' ',7,7' '-hexahydroxy- (7CI,8CI), Ginkgetin, didemethyl- (6CI), 8-[5-(5,7-Dihydroxy-4-oxo-4H-1-benzopyran-2-yl)-2-hydroxyphenyl]-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, Amenthoflavone, Amentoflavone, I3',II8-Biapigenin, NSC 295677, Tridemethylsciadopitysin
IUPAC-namn 8-[5-(5,7-dihydroxy-4-oxochromen-2-yl)-2-hydroxyphenyl]-5,7-dihydroxy-2-(4-hydroxyphenyl)chromen-4-one
LEDER Oc1ccc(cc1)C2=CC(=O)c3c(O)cc(O)c(c3O2)c4cc(ccc4O)C5=CC(=O)c6c(O)cc(O)cc6O5
Specifikationer
Kemiskt namn eller material Amentoflavone
Kvantitet 1 mg
InChI formel InChI=1S/C30H18O10/c31-15-4-1-13(2-5-15)24-12-23(38)29-21(36)10-20(35)27(30(29)40-24)17-7-14(3-6-18(17)33)25-11-22(37)28-19(34)8-16(32)9-26(28)39-25/h1-12,31-36H
Formel vikt 538.09