Learn More
Cytiva Amersham™ Reinforced Nitrocellulose Blotting Membrane
Nitrocellulose membrane, with a 0.45 µm pore size, is suitable for proteins of a wide range of molecular weights.
Brand: Cytiva 10600124
1009.17 SEK valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Additional Details : CAS Number : 9004-70-0 Weight : 0.00700kg
Description
- Low background and excellent signal-to-noise ratios.
- Compatible with a variety of labeling and detection systems.
- High binding capacity.
- Suitable for proteins of a wide range of molecular weights.
- Proprietary nitrocellulose formula gives a long shelf life of bound proteins.
- Available in popular precut sizes for your application.
Specifications
Blotting Membrane | |
Protein | |
1 Each | |
For western, southern and northern blotting applications |
Safety and Handling
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose