Läs mer
Tocris Bioscience™ AMG 517
GREENER_CHOICE
Produktkod. 30001014
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30001014 50 mg
30000812 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30001014 Leverantör Tocris Bioscience™ Leverantörsnummer 5995/50
Potent TRPV1 antagonist
Potent TRPV1 antagonist (IC50 = 0.9 nM). Inhibits capsaican-induced flinching and reverses analgesia in an inflammatory pain model in rats. Orally bioavailable.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 659730-32-2
Molekylformel C20H13F3N4O2S
Specifikationer
Kemiskt namn eller material N-[4-[[6-[4-(Trifluoromethyl)phenyl]-4-pyrimidinyl]oxy]-2-benzothiazolyl]acetamide
Kvantitet 50 mg
Löslighet Soluble to 100mM in DMSO
Formel vikt 430.41
Renhet >98%
Molekylvikt (g/mol) 430.405