Läs mer
Tocris Bioscience™ AMG PERK 44
Potent and selective PERK inhibitor; orally bioavailable
Brand: Tocris Bioscience™ 5517/50
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
AMG PERK 44 is a potent and selective PERK inhibitor (IC50 = 6 nM). Exhibits >160-fold selectivity for PERK over B-Raf and GCN2, as well as a panel of 387 other kinases. Orally bioavailable.Specifikationer
| 4-[2-Amino-4-methyl-3-(2-methyl-6-quinolinyl)benzoyl]-1,2-dihydro-1-methyl-2,5-diphenyl-3H-pyrazol-3-one hydrochloride | |
| 1883548-84-2 | |
| 50 mg |