missing translation for 'onlineSavingsMsg'
Learn More

AMH Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14461-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16823114

  • 3313.85 SEK / 150µL
Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody is specific to AMH.

Anti mullerian hormone (AMH) is a member of the TGF beta superfamily. It is secreted as a homodimeric 140 kDa disulfide linked precursor that is cleaved to release the mature 30 kDa homodimer. Originally classified as a fetal testicular hormone that inhibits Mullerian duct development, AMH is expressed post natally by immature Sertoli cells, and to a lesser degree by granulosa cells. AMH plays a role in testicular differentiation and in the regulation of ovarian follicle growth.
Specifications

Specifications

AMH
0.41 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P03971, P27106
AMH, anti Mullerian hormone, MIF, MIS
Rabbit
IgG
150 μL
-20°C
Primary
11705, 268
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
AMH
Liquid
AMH
AMH Fusion Protein Ag5670
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Videos
SDS
Documents
Certificates
Special Offers

Special Offers