Läs mer
Amicarbalide Diisethionate, LoGiCal
Produktkod. 30457710
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30457710 5 mg
1 options
Produktkod. 30457710 Leverantör LoGiCal Leverantörsnummer LGCFOR1779.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Amicarbalide Diisethionate
CAS 3671-72-5
Analyt- eller komponentnamn Amicarbalide Diisethionate
Frakt skick Room Temperature
Molekylformel C15 H16 N6 O . 2 C2 H6 O4 S
Certifikat och efterlevnad ISO 9001
Synonym Carbanilide, 3,3'-diamidino-, diisethionate (6CI), Ethanesulfonic acid, 2-hydroxy-, compd. with 3,3'-diamidinocarbanilide (2:1) (8CI), Benzenecarboximidamide, 3,3'-(carbonyldiimino)bis-, bis(2-hydroxyethanesulfonate) (9CI), Carbanilide, 3,3'-diamidino-, bis(2-hydroxyethanesulfonate) (8CI), Amicarbalide diisethionate, Amicarbalide isethionate, Diampron, Diamprone
InChI formel InChI=1S/C15H16N6O.2C2H6O4S/c16-13(17)9-3-1-5-11(7-9)20-15(22)21-12-6-2-4-10(8-12)14(18)19;2*3-1-2-7(4,5)6/h1-8H,(H3,16,17)(H3,18,19)(H2,20,21,22);2*3H,1-2H2,(H,4,5,6)
Lösningstyp Neat
LEDER NC(=N)c1cccc(NC(=O)Nc2cccc(c2)C(=N)N)c1.OCCS(=O)(=O)O.OCCS(=O)(=O)O
IUPAC-namn 1,3-bis(3-carbamimidoylphenyl)urea;2-hydroxyethanesulfonic acid
Industry Type Clinical Testing
Molekylvikt (g/mol) 548.59
Kvantitet 5 mg
Formel vikt 548.1359 g/mol