Läs mer
Amidosulfuron, TRC
Produktkod. 30133648
Change view
Kvantitet:
100 mg
500 mg
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133648 10 g
30132996 500 mg
17960405 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133648 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA57679510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 120923-37-7
Molekylformel C9 H15 N5 O7 S2
Molekylvikt (g/mol) 369.37
Synonym 2,4-Dithia-3,5-diazahexan-6-amide, N-(4,6-dimethoxy-2-pyrimidinyl)-3-methyl-, 2,2,4,4-tetraoxide (9CI), Adret, Amidosulfuron, Cratil, Gratil, Grodyl, HOE 075032, Hoestar
IUPAC-namn 1-(4,6-dimethoxypyrimidin-2-yl)-3-[methyl(methylsulfonyl)sulfamoyl]urea
LEDER COc1cc(OC)nc(NC(=O)NS(=O)(=O)N(C)S(=O)(=O)C)n1
Specifikationer
Kemiskt namn eller material Amidosulfuron
Kvantitet 10 g
InChI formel InChI=1S/C9H15N5O7S2/c1-14(22(4,16)17)23(18,19)13-9(15)12-8-10-6(20-2)5-7(11-8)21-3/h5H,1-4H3,(H2,10,11,12,13,15)
Formel vikt 369.0413