Learn More
AMIGO1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561164
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SSVMDFQEDL YCMNSKKLHN VFNLSFLNCG EYKERAWEAH LGDTLIIKCD TKQQGMTKVW VTPSNERVLD EVTNGTVSVS KDG Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 81%.
AMIGO1 Polyclonal antibody specifically detects AMIGO1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
AMIGO1 | |
Polyclonal | |
Unconjugated | |
Amigo1 | |
adhesion molecule with Ig like domain 1; adhesion molecule with Ig-like domain 1; ali2; alivin 2; alivin-2; AMIGO; AMIGO 1; AMIGO1; AMIGO-1; amigo1 {ECO:0000250; amphoretin-induced gene and ORF; amphoterin induced gene and ORF (Amigo); amphoterin-induced gene and ORF; amphoterin-induced protein 1; KIAA1163; MGC25558; OTTHUMP00000013379; RP23 89M15.6; transmembrane protein AMIGO; UniProtKB:Q80ZD8} | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57463 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86WK6 | |
Amigo1 | |
Recombinant protein corresponding to Human AMIGO1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only