missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ε-Amino-N-caproic Acid, 99.8%, MP Biomedicals™
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Specifications
Color | White |
Density | 1.48g/mL |
Assay Percent Range | 99.8% |
Quantity | 500 g |
Formula Weight | 131.2 |
Physical Form | Crystalline Powder |
Chemical Name or Material | ε-Amino-N-caproic Acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'