missing translation for 'onlineSavingsMsg'
Learn More

ε-Amino-N-caproic Acid, 99.8%, MP Biomedicals™

Catalog Number p-4404366
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
This item is not returnable. View return policy
60-32-2
C6H13NO2
131.175
SLXKOJJOQWFEFD-UHFFFAOYSA-N
6-aminocaproic acid, aminocaproic acid, amicar, epsikapron, capramol, caprocid, epsamon, eaca, acepramin, caprolisin
564
CHEBI:16586
6-aminohexanoic acid
C(CCC(=O)O)CCN
This item is not returnable. View return policy
60-32-2
C6H13NO2
131.175
SLXKOJJOQWFEFD-UHFFFAOYSA-N
6-aminocaproic acid, aminocaproic acid, amicar, epsikapron, capramol, caprocid, epsamon, eaca, acepramin, caprolisin
564
CHEBI:16586
6-aminohexanoic acid
C(CCC(=O)O)CCN
Color White
Density 1.48g/mL
Assay Percent Range 99.8%
Quantity 25 g
Formula Weight 131.2
Physical Form Crystalline Powder
Chemical Name or Material ε-Amino-N-caproic Acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'