missing translation for 'onlineSavingsMsg'
Learn More

Aminoacetaldehyde dimethyl acetal, 99%, Thermo Scientific Chemicals

Catalog Number p-56113
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
22483-09-6
MFCD00008135
QKWWDTYDYOFRJL-UHFFFAOYSA-N
aminoacetaldehyde dimethyl acetal, 2,2-dimethoxyethylamine, ethanamine, 2,2-dimethoxy, 2,2-dimethoxyethan-1-amine, dimethylaminoacetal, amino acetaldehyde dimethyl acetal, 1-amino-2,2-dimethoxyethane, 2-aminoacetaldehyde dimethyl acetal, acetaldehyde, amino-, dimethyl acetal, acmc-209fwr
89728
2,2-dimethoxyethanamine
COC(CN)OC
This item is not returnable. View return policy
22483-09-6
MFCD00008135
QKWWDTYDYOFRJL-UHFFFAOYSA-N
aminoacetaldehyde dimethyl acetal, 2,2-dimethoxyethylamine, ethanamine, 2,2-dimethoxy, 2,2-dimethoxyethan-1-amine, dimethylaminoacetal, amino acetaldehyde dimethyl acetal, 1-amino-2,2-dimethoxyethane, 2-aminoacetaldehyde dimethyl acetal, acetaldehyde, amino-, dimethyl acetal, acmc-209fwr
89728
2,2-dimethoxyethanamine
COC(CN)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 0.9700g/mL
Boiling Point 135°C to 139°C (95.0 mmHg)
Flash Point 41°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C4H11NO2
Linear Formula H2NCH2CH(OCH3)2
Refractive Index 1.4160 to 1.418
Quantity 25 g
Beilstein 04, II, 758
Specific Gravity 0.97
Solubility Information Solubility in water: miscible
Molecular Weight (g/mol) 105.14
Viscosity 0.15 Pas (20°C)
Formula Weight 105.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Aminoacetaldehyde dimethyl acetal, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Aminoacetaldehyde dimethyl acetal
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'