Läs mer
(Aminocarbonyl)azanyl Carbamic Acid Ester, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | (Aminocarbonyl)azanyl Carbamic Acid Ester |
| CAS | 4543-62-8 |
| Molekylformel | C2 H5 N3 O3 |
| Kvantitet | 50 mg |
| Synonym | Carbamoyloxyurea, (Aminocarbonyl)azanyl Carbamate, N,O-Dicarbamoyl-hydroxylamine, [(Aminocarbonyl)oxy]-urea, N,O-Bis(aminocarbonyl)-hydroxylamine, N-(Carbamoyloxy)urea |
| InChI formel | InChI=1S/C2H5N3O3/c3-1(6)5-8-2(4)7/h(H2,4,7)(H3,3,5,6) |
| LEDER | NC(=O)NOC(=O)N |
| IUPAC-namn | (carbamoylamino) carbamate |
| Molekylvikt (g/mol) | 119.079 |
| Formel vikt | 119.033 |