missing translation for 'onlineSavingsMsg'
Learn More
Learn More
δ-Aminolevulinic Acid, 98%, MP Biomedicals
Click to view available options
Quantity:
25 mg
500 mg
1 g
Specifications
Quantity | 25 mg |
Synonym | 5-aminolevulinic acid hydrochloride, 5-amino-4-oxopentanoic acid hydrochloride, aminolevulinic acid hydrochloride, levulan, levulan kerastick, aminolevulinic acid hcl, delta-aminolevulinic acid hydrochloride, gliolan, ameluz, 5-aminolevulinate hydrochloride |
SMILES | C(CC(=O)O)C(=O)CN.Cl |
IUPAC Name | 5-amino-4-oxopentanoic acid;hydrochloride |
PubChem CID | 123608 |
Formula Weight | 167.6 |
Grade | Cell Culture Reagent |
Chemical Name or Material | δ-Aminolevulinic Acid, 98%, Hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'