Läs mer
Aminopterin, TRC
Produktkod. 18012510
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18012510 100 mg
18070300 250 mg
18041510 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18012510 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA628850100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 54-62-6
Molekylformel C19 H20 N8 O5
Molekylvikt (g/mol) 440.41
Synonym L-Glutamic acid, N-[4-[[(2,4-diamino-6-pteridinyl)methyl]amino]benzoyl]-, Glutamic acid, N-[p-[[(2,4-diamino-6-pteridinyl)methyl]amino]benzoyl]-, L- (8CI), N-[4-[[(2,4-Diamino-6-pteridinyl)methyl]amino]benzoyl]-L-glutamic acid, 4-Aminofolic acid, 4-Aminopteroylglutamic acid, APGA, Aminopterin, Aminopterine, NSC 739, Pteramina, (S)-2-{4-[(2,4-Diaminopteridin-6-yl)methylamino]benzamido}pentanedioic acid, Methotrexate Imp. B (EP), (2S)-2-[[4-[[(2,4-Diaminopteridin-6-yl)methyl]amino]benzoyl]amino]pentanedioic Acid
IUPAC-namn (2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid
LEDER Nc1nc(N)c2nc(CNc3ccc(cc3)C(=O)N[C@@H](CCC(=O)O)C(=O)O)cnc2n1
Specifikationer
Kemiskt namn eller material Aminopterin
Analyt- eller komponentnamn Aminopterin
Anteckningar om renhetsgrad HPLC
Frakt skick Room Temperature
InChI formel InChI=1S/C19H20N8O5/c20-15-14-16(27-19(21)26-15)23-8-11(24-14)7-22-10-3-1-9(2-4-10)17(30)25-12(18(31)32)5-6-13(28)29/h1-4,8,12,22H,5-7H2,(H,25,30)(H,28,29)(H,31,32)(H4,20,21,23,26,27)/t12-/m0/s1
Lösningstyp Neat
Industry Type Forensics
Kvantitet 100 mg
Formel vikt 440.1557
Procent renhet >95%