Läs mer
Amiodarone EP Impurity D, Mikromol™
Produktkod. 17991176
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17991176 25 mg
17942156 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17991176 Leverantör Mikromol Leverantörsnummer MM0697.050025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Amiodarone EP Impurity D
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2-Butylbenzofuran-3-Yl)(4-Hydroxy-3,5-Diiodophenyl)Methanone
CAS 1951-26-4
Analyt- eller komponentnamn (2-Butylbenzofuran-3-Yl)(4-Hydroxy-3,5-Diiodophenyl)Methanone
Frakt skick Room Temperature
Molekylformel C19 H16 I2 O3
Certifikat och efterlevnad ISO 17025
Synonym (2-Butylbenzofuran-3-yl)(4-hydroxy-3,5-diiodophenyl)methanone, Amiodarone Hydrochloride Imp. D (EP), Amiodarone Imp. D (EP), Amiodarone USP Related Compound D, Amiodarone USP RC D, (2-Butylbenzofuran-3-yl)(4-hydroxy-3,5-diiodophenyl)methanone, Amiodarone Related Compound D, Amiodarone Hydrochloride Impurity D, Amiodarone Impurity D
InChI formel InChI=1S/C19H16I2O3/c1-2-3-7-16-17(12-6-4-5-8-15(12)24-16)18(22)11-9-13(20)19(23)14(21)10-11/h4-6,8-10,23H,2-3,7H2,1H3
LEDER CCCCc1oc2ccccc2c1C(=O)c3cc(I)c(O)c(I)c3
IUPAC-namn (2-butyl-1-benzofuran-3-yl)-(4-hydroxy-3,5-diiodophenyl)methanone
Molekylvikt (g/mol) 546.14
Kvantitet 25 mg
Formel vikt 545.9189 g/mol