Läs mer
Amiquinsin Hydrochloride Monohydrate, LoGiCal
Produktkod. 30463700
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30463700 10 mg
1 options
Produktkod. 30463700 Leverantör LoGiCal Leverantörsnummer LGCFOR3280.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Amiquinsin Hydrochloride Monohydrate
CAS 7125-70-4
Analyt- eller komponentnamn Amiquinsin Hydrochloride Monohydrate
Frakt skick Room Temperature
Molekylformel C11 H12 N2 O2 . Cl H . H2 O
Certifikat och efterlevnad ISO 9001
Synonym 4-Quinolinamine, 6,7-dimethoxy-, hydrochloride, hydrate (1:1:1), 4-Quinolinamine, 6,7-dimethoxy-, monohydrochloride, monohydrate (9CI), Quinoline, 4-amino-6,7-dimethoxy-, monohydrochloride, monohydrate (8CI), 4-Amino-6,7-dimethoxyquinoline hydrochloride monohydrate, 6,7-Dimethoxy-4-aminoquinoline hydrochloride hydrate, Amiquinsin hydrochloride, U 935, U 936
InChI formel InChI=1S/C11H12N2O2.ClH.H2O/c1-14-10-5-7-8(12)3-4-13-9(7)6-11(10)15-2;;/h3-6H,1-2H3,(H2,12,13);1H;1H2
Lösningstyp Neat
LEDER O.Cl.COc1cc2nccc(N)c2cc1OC
IUPAC-namn 6,7-dimethoxyquinolin-4-amine;hydrate;hydrochloride
Industry Type Clinical Testing
Molekylvikt (g/mol) 258.7
Kvantitet 10 mg
Formel vikt 258.0771 g/mol