Läs mer
Amiquinsin Hydrochloride Monohydrate, Mikromol™
Produktkod. 30049730
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30049730 50 mg
1 options
Produktkod. 30049730 Leverantör Mikromol Leverantörsnummer MM3280.00
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Amiquinsin Hydrochloride Monohydrate
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Amiquinsin Hydrochloride Monohydrate
CAS 7125-70-4
Analyt- eller komponentnamn Amiquinsin Hydrochloride Monohydrate
Frakt skick Room Temperature
Molekylformel C11 H12 N2 O2 . Cl H . H2 O
Certifikat och efterlevnad ISO 17025
Synonym 4-Quinolinamine, 6,7-dimethoxy-, hydrochloride, hydrate (1:1:1), 4-Quinolinamine, 6,7-dimethoxy-, monohydrochloride, monohydrate (9CI), Quinoline, 4-amino-6,7-dimethoxy-, monohydrochloride, monohydrate (8CI), 4-Amino-6,7-dimethoxyquinoline hydrochloride monohydrate, 6,7-Dimethoxy-4-aminoquinoline hydrochloride hydrate, Amiquinsin hydrochloride, U 935, U 936
InChI formel InChI=1S/C11H12N2O2.ClH.H2O/c1-14-10-5-7-8(12)3-4-13-9(7)6-11(10)15-2;;/h3-6H,1-2H3,(H2,12,13);1H;1H2
LEDER O.Cl.COc1cc2nccc(N)c2cc1OC
IUPAC-namn 6,7-dimethoxyquinolin-4-amine;hydrate;hydrochloride
Molekylvikt (g/mol) 258.7
Kvantitet 50 mg
Formel vikt 258.0771 g/mol