Beskrivning
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Specifikationer
Specifikationer
| Kemiskt namn eller material | Amitraz |
| Förpackning | Glass Bottle |
| CAS | 33089-61-1 |
| Färg | White/Yellow |
| Smältpunkt | 86°C to 88°C |
| Synonym | N′-(2,4-Dimethylphenyl)-N-{[(2,4-dimethylphenyl)imino]methyl}-N-methylmethanimidamide |
| Industry Type | Environmental Analysis |
| Molekylvikt (g/mol) | 293.4 |
| Kvantitet | 100 mg |
| Kvalitet | Analytical Standard |