Läs mer
Amitriptyline N-Oxide Hydrochloride, Mikromol™
Beskrivning
Amitriptyline N-Oxide Hydrochloride
Specifikationer
Specifikationer
| Kemiskt namn eller material | Amitriptyline N-Oxide Hydrochloride |
| CAS | 4290-60-2 |
| Analyt- eller komponentnamn | Amitriptyline N-Oxide Hydrochloride |
| Frakt skick | Room Temperature |
| Molekylformel | C20 H23 N O . Cl H |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | Nortriptyline Hydrochloride Imp. J (EP), Nortriptyline Imp. J (EP), [3-(10,11-Dihydro-5H-dibenzo[a,d][7]annulen-5-ylidene)propyl]-dimethylamine Oxide Hydrochloride, Amitriptyline N-Oxide Hydrochloride, Nortriptyline Hydrochloride Imp. J (EP) as Hydrochloride, Nortriptyline Hydrochloride Impurity J as Hydrochloride, Nortriptyline Impurity J as Hydrochloride |
| InChI formel | InChI=1S/C20H23NO.ClH/c1-21(2,22)15-7-12-20-18-10-5-3-8-16(18)13-14-17-9-4-6-11-19(17)20;/h3-6,8-12H,7,13-15H2,1-2H3;1H |
| LEDER | Cl.C[N+](C)([O-])CCC=C1c2ccccc2CCc3ccccc13 |
| IUPAC-namn | 3-(5,6-dihydrodibenzo[2,1-b:1',2'-e][7]annulen-11-ylidene)-N,N-dimethylpropan-1-amine oxide;hydrochloride |
| Visa mer |