Läs mer
Amitriptyline, TRC
Produktkod. 30329708
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30329708 100 mg
1 options
Produktkod. 30329708 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA640860100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Amitriptyline
CAS 50-48-6
Anteckningar om renhetsgrad HPLC
Molekylformel C20 H23 N
Kvantitet 100 mg
Synonym 1-Propanamine, 3-(10,11-dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-N,N-dimethyl-, 5H-Dibenzo[a,d]cycloheptene-δ5,γ-propylamine, 10,11-dihydro-N,N-dimethyl- (6CI,8CI), 3-(10,11-Dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-N,N-dimethyl-1-propanamine, 5-(3-Dimethylaminopropylidene)-5H-dibenzo[a,d]-10,11-dihydrocycloheptene, 5-(3'-Dimethylaminopropylidene)-dibenzo-[a,d][1,4]-cycloheptadiene, 5-(γ-Dimethylaminopropylidene)-10,11-dihydro-5H-dibenzo[a,d]cycloheptene, 5-(γ-Dimethylaminopropylidene)-5H-dibenzo(a,d)(1,4)cycloheptadiene, Adepress, Amitriprolidine, Amitriptylin, Amitriptyline, Damilen, Damitriptyline, Flavyl, MK 230, N 750, Proheptadiene, Seroten, Triptanol, Triptisol
InChI formel InChI=1S/C20H23N/c1-21(2)15-7-12-20-18-10-5-3-8-16(18)13-14-17-9-4-6-11-19(17)20/h3-6,8-12H,7,13-15H2,1-2H3
LEDER CN(C)CCC=C1c2ccccc2CCc3ccccc13
IUPAC-namn 3-(5,6-dihydrodibenzo[2,1-b:1',2'-e][7]annulen-11-ylidene)-N,N-dimethylpropan-1-amine
Molekylvikt (g/mol) 277.4033
Formel vikt 277.183