Läs mer
Invitrogen™ Amlitelimab Recombinant Monoclonal Antibody
Produktkod. 30284210
Change view
Kvantitet:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30284210 100 μg
30282548 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30284210 Leverantör Invitrogen™ Leverantörsnummer MA559061
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Recombinant Monoclonal Antibody
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Amlitelimab is a monoclonal antibody that targets and inhibits OX40L, a protein involved in the activation and survival of T cells, thereby modulating the immune response.
TRUSTED_SUSTAINABILITY
Specifikationer
Antigen Amlitelimab
Användningsområden ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Klassificering Recombinant Monoclonal
Koncentration 1 mg/mL
Konjugera Unconjugated
Formulering 25mM Histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Värd art Human
Reningsmetod Protein A
Kvantitet 100 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Målarter Human
Innehåll och lagring -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Lyophilized
Isotyp IgG4SP