Läs mer
Amlodipine Diethyl Ester Maleate, TRC
Produktkod. 30333382
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30333382 100 mg
1 options
Produktkod. 30333382 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA633530100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Amlodipine Diethyl Ester Maleate
CAS 400024-12-6
Molekylformel C21 H27 Cl N2 O5 . C4 H4 O4
Kvantitet 100 mg
Synonym Diethyl 2-((2-Aminoethoxy)methyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate Maleate, 2-[(2-Aminoethoxy)methyl]-4-(2-chlorophenyl)-1,4-dihydro-6-methyl-3,5-pyridinedicarboxylic Acid Diethyl Ester (2Z)-2-Butenedioate (1:1)
InChI formel InChI=1S/C21H27ClN2O5.C4H4O4/c1-4-28-20(25)17-13(3)24-16(12-27-11-10-23)19(21(26)29-5-2)18(17)14-8-6-7-9-15(14)22;5-3(6)1-2-4(7)8/h6-9,18,24H,4-5,10-12,23H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1-
LEDER CCOC(=O)C1=C(C)NC(=C(C1c2ccccc2Cl)C(=O)OCC)COCCN.OC(=O)\C=C/C(=O)O
IUPAC-namn (Z)-but-2-enedioic acid;diethyl 2-(2-aminoethoxymethyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
Molekylvikt (g/mol) 538.975
Formel vikt 538.172