Läs mer
Amlodipine Maleate Adduct, TRC
Produktkod. 30326304
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30326304 10 mg
1 options
Produktkod. 30326304 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA57524010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Amlodipine aspartate
CAS 400602-35-9
Anteckningar om renhetsgrad HPLC
Molekylformel C24 H29 Cl N2 O9
Kvantitet 10 mg
Synonym 4-(2-Chlorophenyl)-2-[[2-[(1,2-dicarboxyethyl)amino]ethoxy]methyl]-1,4-dihydro-6-methyl-3,5-Pyridinedicarboxylic Acid 3-Ethyl 5-Methyl Ester, Amlodipine aspartate, L-Aspartate amlodipine, Amlodipine Aspartic Acid Impurity
InChI formel InChI=1S/C24H29ClN2O9/c1-4-36-24(33)21-17(12-35-10-9-26-16(22(30)31)11-18(28)29)27-13(2)19(23(32)34-3)20(21)14-7-5-6-8-15(14)25/h5-8,16,20,26-27H,4,9-12H2,1-3H3,(H,28,29)(H,30,31)
LEDER O=C(OC)C1=C(NC(=C(C(=O)OCC)C1C=2C=CC=CC2Cl)COCCNC(C(=O)O)CC(=O)O)C
IUPAC-namn 2-[2-[[4-(2-chlorophenyl)-3-ethoxycarbonyl-5-methoxycarbonyl-6-methyl-1,4-dihydropyridin-2-yl]methoxy]ethylamino]butanedioic acid
Molekylvikt (g/mol) 524.95
Formel vikt 524.1562