Läs mer
Ammeline-13C3 Hydrochloride, TRC
Produktkod. 30134110
Change view
Kvantitet:
0.5 mg
5 mg
Förpackningsstorlek:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30134110 5 mg -
30005406 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134110 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6336625MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1794936-94-9
Molekylformel 13C3 H5 N5 O . Cl H
Molekylvikt (g/mol) 166.54
Synonym 4,6-Diamino-1,3,5-triazin-2(1H)-one-13C3 Hydrochloride, 2,4-Diamino-6-hydroxy-1,3,5-triazine-13C3 Hydrochloride, 2,4-Diamino-6-hydroxy-s-triazine-13C3 Hydrochloride, 2-Hydroxy-4,6-diamino-s-triazine-13C3 Hydrochloride, Ammelin-13C3 Hydrochloride, Deethyldeisopropylhydroxyyatrazine-13C3 Hydrochloride, GS 11791-13C3 Hydrochloride, NSC 9778-13C3 Hydrochlorides, Diaminohydroxytriazine-13C3 Hydrochloride
IUPAC-namn 2,6-diamino-(2,4,6-^{13}C_{3})1H-1,3,5-triazin-4-one;hydrochloride
LEDER Cl.N[13C]1=N[13C](=O)N=[13C](N)N1
Specifikationer
Kemiskt namn eller material Ammeline-13C3 Hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C3H5N5O.ClH/c4-1-6-2(5)8-3(9)7-1;/h(H5,4,5,6,7,8,9);1H/i1+1,2+1,3+1;
Formel vikt 166.036
Procent renhet >95%