missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonia, 25% w/w, Reagent Grade, Scharlab
Click to view available options
Quantity:
1 L
2.5 L
Description
- Synonyms: Ammonia water, Ammonium hydroxide solution
- Molecular Formula: NH3
- Molar Mass: 17.03g/mol
- CAS: 1336-21-6
- EIN ECS No.: 215-647-6
Specifications
Boiling Point | 37.7°C |
Melting Point | -57.5°C |
Color | Colorless |
Physical Form | Liquid |
Quantity | 1 L |
Formula Weight | 17.03g/mol |
Percent Purity | 25 to 30% (acidimetric, NH3) |
Grade | Reagent |
Vapor Pressure | ∼500hPa at 20°C |
Density | 0.90g/cm3 |
Safety and Handling
Product Identifier
- Ammonia, solution 25% w/w, reagent grade, Reag. Ph Eur
- Danger
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- Specific target organ toxicity Category 3
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H400-Very toxic to aquatic life.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : Ammonia
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'