Läs mer
Hanna Instruments™ Ammonia Portable Photometer Kit
Typ:
Photometer
Förpackningsstorlek:
Varje
Beskrivning
- Combination of light emitting diode (LED), narrow band interference filter, and silicon photodetector ensure accurate photometric readings every time
- CAL Checktrade; feature utilizes ready-made standards traceable to NIST Standard for performance verification and calibration
- Exclusive cuvette locking system ensures that cuvette is inserted into measurement cell in same position every time to maintain consistent path length
- Combines accuracy and ease of use in ergonomic, portable design
Specifikationer
Specifikationer
| Typ | Photometer |
| Noggrannhet | ±0.05mg/L ±5% of reading |
| För användning med (applikation) | Measurement of ammonia (as NH3-N) in freshwater samples |
| Inkluderar | Portable Photometer, Two sample cuvettes, Two Caps, CAL Check standards with certificate, Cuvette cleaning cloth, Rugged carrying case, Thermoformed insert for meter and accessories |
| Ljuskälla | LED |
| Räckvidd | 0.00 to 9.99mg/L (ppm) (as NH₃−N) |
| Upplösning | 0.01 mg/L |
| Vikt (metrisk) | 360 g |