Läs mer
Ammonium 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Heptadecafluorooctane-1-sulfonate (Technical Grade), TRC
Product Code. 30310535
Change view
Click to view available options
Kvantitet:
100 mg
Product Code. Kvantitet
30310535 100 mg
1 options
Product Code. 30310535 Supplier Toronto Research Chemicals Supplier No. TRCA301920100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material Ammonium 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Heptadecafluorooctane-1-sulfonate
CAS 29081-56-9
Molekylformel C8H4F17NO3S
Kvantitet 100 mg
Synonym 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-1-Octanesulfonic acid ammonium salt (1:1), 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-1-Octanesulfonic acid ammonium salt (8CI,9CI), Ammonium perfluorooctylsulfonate, EF 104, Eftop EF 104, FC 93, Fluorad FC 93, Perfluorooctanesulfonic acid, ammonium salt
InChI formel InChI=1S/C8HF17O3S.H3N/c9-1(10,3(13,14)5(17,18)7(21,22)23)2(11,12)4(15,16)6(19,20)8(24,25)29(26,27)28;/h(H,26,27,28);1H3
LEDER N.O[S](=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC-namn azanium;1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane-1-sulfonate
Molekylvikt (g/mol) 517.16
Formel vikt 516.964