Läs mer
Ammonium-15N Chloride, TRC
Produktkod. 30011915
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011915 1 g
30011837 10 g
30005273 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011915 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6338521G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 39466-62-1
Molekylformel Cl . H4 15N
Molekylvikt (g/mol) 54.48
Synonym Ammonium-15N Chloride
IUPAC-namn azanium;chloride
LEDER [15NH4+].[Cl-]
Specifikationer
Kemiskt namn eller material Ammonium-15N Chloride
Kvantitet 1 g
InChI formel InChI=1S/ClH.H3N/h1H;1H3/i;1+1
Formel vikt 54.0003