Learn More
Ammonium acetate, 97+%, MP Biomedicals™
Click to view available options
Quantity:
100 g
500 g
Specifications
Color | White |
Physical Form | Powder or Liquid |
Quantity | 100 g |
Assay Percent Range | 0.97+ |
Formula Weight | 77.08 |
Density | 1.17 (water = 1) |
Chemical Name or Material | Ammonium acetate |