Learn More
Ammonium Carbonate, Supelco™
Product Code. 15858110
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
Product Code. Quantity Unit Size
15858110 1 kg 1kg
1 options
MQ300 Graded - For Products Used in Applications Requiring Enhanced Change Control and Quality Agreement
Product Code. 15858110 Supplier Supelco Supplier No. 1595041000
 Restricted Item
Beräknad leverans: 26-11-2026
MQ300 Graded - For Products Used in Applications Requiring Enhanced Change Control and Quality Agreement
For analysis EMSURE™ ACS,Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Specifications
CAS 10361-29-2
Quantity 1 kg
MDL Number MFCD00010890
Synonym Ammonium carbonate, salt of hartshorn, Ammonium carbonate
Grade Research
Chemical Name or Material Ammonium Carbonate