missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium cerium(IV) nitrate, 99%, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
50 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Melting Point | 107.0°C to 108.0°C |
Color | Beige-Yellow to White |
Physical Form | Flakes or Powder |
Quantity | 25 g |
Assay Percent Range | 99% |
Linear Formula | (NH4)2Ce(NO3)6 |
Fieser | 01,120; 02,63; 03,44; 04,71; 05,101; 06,99; 09,99; 10,79; 15,70; 16,66; 17,68 |
Merck Index | 15,1991 |
Solubility Information | Solubility in water: 1410g/L (20°C). Other solubilities: insoluble in fat, soluble in alcohol |
Formula Weight | 548.22 |
Show More |
Safety and Handling
Product Identifier
- Ammonium cerium(IV) nitrate
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Oxidising solid Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 C
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H290-May be corrosive to metals.
- H272-May intensify fire; oxidiser.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'