Läs mer
Ammonium chloride, 0.5M aq. soln., Thermo Scientific™
Produktkod. 15404509
Kvantitet:
50 mL
100 mL
Förpackningsstorlek:
100ml
50ml
Produktkod. 15404509
Brand: Thermo Scientific Alfa Aesar J62046.AD
Request Bulk or Custom Format
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Ammonium chloride is used for the lysis of human red blood cells. It is used in protein isolation techniques and in the study of basic calcium phosphate crystals in fibroblasts. It inhibits intracellular dissolution of basic calcium phosphate (BCP) crystals by alkalinizing phagolysosomes. It is used as a flavoring agent in some types of liquorices. It is also used as electrolyte in zinc-carbon batteries.
Solubility
Fully miscible in water. Soluble in ethanol, methanol, and glycerol. Insoluble in acetone, and ether.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 12125-02-9
Molekylformel ClH4N
Molekylvikt (g/mol) 53.49
MDL-nummer MFCD00011420
InChI-nyckel NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC-namn azanium;chloride
LEDER N.Cl
Specifikationer
Kemiskt namn eller material Ammonium chloride
Färg Colorless
Fysisk form Liquid
Kvantitet 50 mL
Linjär formel NH4Cl
Beilstein 4371014
Löslighetsinformation Fully miscible in water. Soluble in ethanol,methanol,and glycerol. Insoluble in acetone,and ether.
Formel vikt 53.49
Koncentration eller sammansättning (efter analyt eller komponenter) 0.5M aq. soln.