Learn More
Ammonium chloride 99.5-100.5% (dried basis), granular USP, FCC, Macron Fine Chemicals™
Brand: Macron Fine Chemicals 3364-12
Additional Details : CAS Number : 12125-02-9 Weight : 0.50000kg
Description
- Food GMP Manufactured Product
- GMP Manufactured Product
- Meets FCC Requirements
- Meets USP Requirements
- Bulk Food Chemical
- Bulk Pharmaceutical Chemical
- No Class 1,2,3 or other solvents are used/produced in the manufacturing/purification of the product
- Metallic Residues: No metal catalysts or metal reagents, as defined by
- EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of this material
Specifications
12125-02-9 | |
ClH4N | |
NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
ammonium chloride | |
1.5256 g/cm³ (20°C) | |
≤ 0.1% | |
≤ 0.5% | |
4.6 to 6.0 | |
335°C |
99.5 to 100.5% | |
MFCD00011420 | |
[NH4+].[Cl-] | |
53.49 | |
Passes Test | |
≤ 4 mg/kg | |
Mall polystormor poly/plastic bottle | |
Passes Test | |
500 g |