missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Fluoride, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
6 x 500 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 100 g |
Linear Formula | NH4F |
UN Number | 2505 |
Formula Weight | 37.04g/mol |
Percent Purity | ≥98% |
Grade | ACS Reagent |
Packaging | Plastic Bottle |
Residue after Ignition | ≤0.01% |
Chemical Name or Material | Ammonium Fluoride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'